Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-methoxypyridine, 90%, tech.

Catalog No. AC103951000
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 6628-77-9 | C6H8N2O | 124.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6628-77-9
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.14
MDL Number MFCD00006264
InChI Key UUVDJIWRSIJEBS-UHFFFAOYSA-N
Synonym 5-amino-2-methoxypyridine, 2-methoxy-5-aminopyridine, 6-methoxy-3-pyridylamine, 3-pyridinamine, 6-methoxy, 2-methoxy-5-amino pyridine, 6-methoxy-pyridin-3-ylamine, pyridine, 5-amino-2-methoxy, 6-methoxy-3-pyridinylamine, 6-methoxy-3-pyridinamine
PubChem CID 81121
IUPAC Name 6-methoxypyridin-3-amine
SMILES COC1=NC=C(C=C1)N

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 29.0°C to 31.0°C
Color Brown-Yellow or Red
Density 1.5750g/mL
Boiling Point 85.0°C to 90.0°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula CH3O(C5H3N)NH2
Quantity 100 g
Beilstein 22,II,408
Specific Gravity 1.575
Solubility Information Solubility in water: slightly soluble in water. Other solubilities: soluble in methanol, ethanol
Formula Weight 124.14
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material 5-Amino-2-methoxypyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 229-612-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.