Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoethanol, 97%

Catalog No. AC106921000
Click to view available options
Quantity:
5 mL
100 mL
250 mL
1 L
Packaging:
Glass bottle

CAS: 540-51-2 | C2H5BrO | 124.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-51-2
Molecular Formula C2H5BrO
Molecular Weight (g/mol) 124.96
MDL Number MFCD00002827
InChI Key LDLCZOVUSADOIV-UHFFFAOYSA-N
Synonym ethylene bromohydrin, ethanol, 2-bromo, 2-bromoethan-1-ol, glycol bromohydrin, 2-hydroxyethyl bromide, 2-bromo-1-ethanol, 1-bromo-2-ethanol, ethylenebromohydrin, 2-bromoethyl alcohol, 2-bromo-ethanol
PubChem CID 10898
IUPAC Name 2-bromoethanol
SMILES C(CBr)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -80.0°C
Color Colorless to Yellow
Density 1.7600g/mL
Boiling Point 149.0°C to 150.0°C (750.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4900 to 1.4940
Linear Formula BrCH2CH2OH
Quantity 100 mL
Beilstein 01, 338
Fieser 11,77
Specific Gravity 1.76
Merck Index 15, 3846
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with ethanol, ether, soluble in most common organic solvents,, except petroleum ether
Formula Weight 124.96
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Bromoethanol, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water
EINECSNumber 208-748-1
RTECSNumber KJ8225000
TSCA TSCA
Recommended Storage Product may darken during storage

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.