Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromohexane, 99+%

Catalog No. AC107040100
Click to view available options
Quantity:
500 mL
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum

CAS: 111-25-1 | C6H13Br | 165.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-25-1
Molecular Formula C6H13Br
Molecular Weight (g/mol) 165.07
MDL Number MFCD00000271
InChI Key MNDIARAMWBIKFW-UHFFFAOYSA-N
Synonym hexyl bromide, n-hexyl bromide, bromohexane, 1-hexyl bromide, hexane, 1-bromo, hexane, bromo, unii-wva0fax7ga, 1-bromo-hexane, pentane, bromomethyl, wva0fax7ga
PubChem CID 8101
IUPAC Name 1-bromohexane
SMILES CCCCCCBr

Specifications

Acidity 0.1% max. (as HBr)
CAS Min % 99.0
CAS Max % 100.0
Melting Point -85°C
Density 1.1700g/mL
Boiling Point 154°C to 158°C
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 0.5% max. Hexanol (GC)
Packaging Plastic drum
Refractive Index 1.4465 to 1.4485
Linear Formula CH3(CH2)5Br
Quantity 10 L
Beilstein 01,144
Specific Gravity 1.17
Solubility Information Solubility in water: insoluble
Formula Weight 165.07
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 1-Bromohexane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-850-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.