Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyric acid, 99+%

Catalog No. AC108110025
Click to view available options
Quantity:
100 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum

CAS: 107-92-6 | C4H8O2 | 88.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-92-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00002814
InChI Key FERIUCNNQQJTOY-UHFFFAOYSA-N
Synonym butyric acid, n-butyric acid, n-butanoic acid, propylformic acid, ethylacetic acid, butanic acid, 1-propanecarboxylic acid, butyrate, 1-butyric acid, buttersaeure
PubChem CID 264
ChEBI CHEBI:30772
IUPAC Name butanoic acid
SMILES CCCC(O)=O

Specifications

Melting Point -7.0°C to +5.0°C
Boiling Point 162.0°C to 165.0°C
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula CH3CH2CH2CO2H
UN Number 2820
Solubility Information Solubility in water: miscible. Other solubilities: miscible with almost all common organic solvents
Formula Weight 88.11
Percent Purity ≥99%
Density 0.9600g/mL
Quantity 2.5 L
Flash Point 69°C
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.3965 to 1.3995
Specific Gravity 0.96
Chemical Name or Material n-Butyric acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.