Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-2-methylpropane, 99%

Catalog No. AC109505000
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Chloro-2-methylpropane, 99%, C4H9Cl, CAS Number-507-20-0, 2-methyl-2-chloropropane, propane, 2-chloro-2-methyl, 2-chloroisobutane, tert-butylchloride, t-butylchloride, chlorotrimethylmethane, trimethylchloromethane, tert-butyl chloride, t-butyl chloride, 2-methyl-2-propyl chloride, 500mL | CAS: 507-20-0 | C4H9Cl | 92.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 507-20-0
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.57
MDL Number MFCD00000816
InChI Key NBRKLOOSMBRFMH-UHFFFAOYSA-N
Synonym tert-butyl chloride, propane, 2-chloro-2-methyl, tert-butylchloride, chlorotrimethylmethane, trimethylchloromethane, 2-chloroisobutane, t-butyl chloride, t-butylchloride, 2-methyl-2-chloropropane, 2-methyl-2-propyl chloride
PubChem CID 10486
IUPAC Name 2-chloro-2-methylpropane
SMILES CC(C)(C)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -25.0°C
Density 0.8500g/mL
Boiling Point 51.0°C to 52.0°C
Flash Point −27°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3838 to 1.3858
Linear Formula (CH3)3CCl
Quantity 500 mL
Beilstein 01, 125
Fieser 02,46
Specific Gravity 0.85
Merck Index 15, 1563
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with alcohol and ether
Viscosity 0.51 mPa.s (20°C)
Formula Weight 92.57
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Chloro-2-methylpropane, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep container tightly closed.
EINECSNumber 208-066-4
RTECSNumber TX5040000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.