Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl phthalate, 99%

Catalog No. AC114520100
Click to view available options
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum

CAS: 84-66-2 | C12H14O4 | 222.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-66-2
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00009111
InChI Key FLKPEMZONWLCSK-UHFFFAOYSA-N
Synonym diethyl phthalate, phthalic acid diethyl ester, ethyl phthalate, anozol, neantine, phthalol, solvanol, diethyl o-phthalate, palatinol a, placidol e
PubChem CID 6781
ChEBI CHEBI:34698
IUPAC Name diethyl benzene-1,2-dicarboxylate
SMILES CCOC(=O)C1=CC=CC=C1C(=O)OCC

Specifications

Density 1.1180g/mL
Boiling Point 298.0°C to 299.0°C
Flash Point 156°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic drum
Refractive Index 1.5010 to 1.5030
Quantity 10 L
Beilstein 09,798
Merck Index 15,7483
Specific Gravity 1.118
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: soluble in ethanol,diethyl ether,acetone,,benzene,chloroform,carbon tetrachloride
Viscosity 12 mPa.s (20°C)
Formula Weight 222.24
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diethyl phthalate, 99%
Show More Show Less
EINECSNumber 201-550-6
RTECSNumber TI1050000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.