Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indole-2-carboxylic acid, 99%

Catalog No. AC122200100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC122200100 10 g Glass bottle
AC122200500 50 g Glass bottle
2 options

Catalog No. AC122200100

Supplier: Thermo Scientific Chemicals 122200100

Only null left
Add to Cart

CAS: 1477-50-5 | C9H7NO2 | 161.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1477-50-5
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00005611
InChI Key HCUARRIEZVDMPT-UHFFFAOYSA-N
Synonym indole-2-carboxylic acid, 2-carboxyindole, 2-indolecarboxylic acid, indol-2-carboxylic acid, 1h-indolecarboxylic acid, indole-2-carboxylate, 2-indolylformic acid, 2-indole carboxylic acid, 1h-indol-2-carbons, carboxyindole
PubChem CID 72899
IUPAC Name 1H-indole-2-carboxylic acid
SMILES OC(=O)C1=CC2=CC=CC=C2N1

Specifications

Melting Point 205.0°C to 209.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Beilstein 22, 61
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ether
Formula Weight 161.16
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Indole-2-carboxylic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-030-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.