Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrobromic acid, pure, ca. 48 wt% solution in water
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent

Specifications
Specifications
Color | Brown to Yellow |
Melting Point | -11.0°C |
Boiling Point | 126.0°C to 128.0°C |
Density | 1.4800g/mL |
Packaging | Glass bottle |
Specific Gravity | 1.48 |
Quantity | 2.5 L |
Physical Form | Solution |
Linear Formula | HBr |
Fieser | 01,450; 02,214; 03,154; 04,249; 05,332; 06,282; 10,200; 15,273 |
Show More |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement May be corrosive to metals. Causes severe skin burns and eye damage. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. |
EINECSNumber | 233-113-0 |
RTECSNumber | MW3850000 |
TSCA | TSCA |
Recommended Storage | Slowly darkens on exposure to air and light |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction