Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium chloride, 99+%, pure

Catalog No. AC123340251
Click to view available options
Quantity:
25 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic Bottle
Plastic drum

Ammonium chloride, 99+%, H4ClN, CAS Number-12125-02-9, amchlor, ammoneric, ammoniumchlorid, ammonium chloride, ammonium chloride nh4 cl, ammoniumklorid, ammonium muriate, darammon, sal ammoniac, salmiac, 1kg, 99% min. (Argentometry), CHEBI:31206, White, 1.5200g/mL, 235-186-4, 15,13, 53.49, Pure | CAS: 12125-02-9 | ClH4N | 53.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
SMILES N.Cl

Specifications

Melting Point 340.0°C
Color White
Physical Form Crystalline Powder and/or Chunks
Quantity 25 kg
Assay Percent Range 99% min. (Argentometry)
Linear Formula NH3·HCl
Fieser 15,13
Merck Index 15, 506
Solubility Information Solubility in water: 370g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol,practically insoluble in acetone,ether and,ethyl acetate
IUPAC Name amine hydrochloride
Formula Weight 53.49
Percent Purity ≥99%
Grade Pure
Packaging Fibre drum
Density 1.5200g/mL
Chemical Name or Material Ammonium chloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not eat,d
EINECSNumber 235-186-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.