Learn More
Thermo Scientific Chemicals N-Methyl-D-glucamine, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Information
• N-Methyl-D-glucamine is a hexosamine compound that is frequently used in combination with iodinated organic compounds in contrast media such as iothalamate meglumine, iodipamide meglumine, and diatrizoate meglumine
Applications
• It is also useful when applied in ion-exchange resins to chelate boron ions selectively
• It is considered a base and frequently used in the preparation of salts of certain acids
• It is also applied in the synthesis of surface-active agents and laboratory dyes

Specifications
Specifications
Absorbance | 0.03 max. (20 w/v% aq. soln.) at 420nm |
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 128.0°C to 132.0°C |
Color | White |
Loss on Drying | 1% max. (1 g, 100°C, 3 hrs) |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Plastic drum |
Quantity | 2.5 kg |
Show More |
Safety and Handling
Hazard Category | HYGROSCOPIC |
Hazard Statement | AIR SENSITIVE |
EINECSNumber | 228-506-9 |
TSCA | TSCA |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.