Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl pyruvate, 94%

Catalog No. AC127651000
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 600-22-6 | C4H6O3 | 102.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 600-22-6
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
MDL Number MFCD00008754
InChI Key CWKLZLBVOJRSOM-UHFFFAOYSA-N
Synonym methyl pyruvate, pyruvic acid, methyl ester, pyruvic acid methyl ester, methyl 2-oxopropionate, propanoic acid, 2-oxo-, methyl ester, methyl-pyruvate, methylglyoxylic acid methyl ester, unii-3kjm65g5xl, 3kjm65g5xl, 2-oxo-propionic acid methyl ester
PubChem CID 11748
ChEBI CHEBI:51850
IUPAC Name methyl 2-oxopropanoate
SMILES CC(=O)C(=O)OC

Specifications

Color Colorless to Yellow
Density 1.1300g/mL
Boiling Point 134.0°C to 137.0°C
Flash Point 39°C
Infrared Spectrum Authentic
Assay Percent Range 94%
Packaging Glass bottle
Refractive Index 1.4065
Linear Formula CH3COCO2CH3
Quantity 100 g
Beilstein 03, 616
Specific Gravity 1.13
Merck Index 15, 8135
Solubility Information Solubility in water: soluble.
Formula Weight 102.09
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Methyl pyruvate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-987-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.