Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl sulfoxide, 99.7%, pure, Thermo Scientific Chemicals

Catalog No. AC127790250
Click to view available options
Quantity:
100 mL
1 L
2.5 L
5 L
25 L
Packaging:
Glass Bottle
Plastic Drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methylsulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Density 1.1000g/mL
Boiling Point 189.0°C
Quantity 25 L
Flash Point 87°C
Infrared Spectrum Authentic
Packaging Plastic Drum
Physical Form Liquid
Refractive Index 1.4770 to 1.4800
Assay Percent Range 99.7%
Specific Gravity 1.1
Linear Formula (CH3)2SO
Viscosity 1.987 mPa.s (25°C)
Water 0.2% max.
Fieser 01,296; 02,157; 03,119; 04,192; 05,263; 06,225; 07,133; 08,198; 09,189; 10,166; 12,212; 13,124; 15,146; 16,149; 17,127
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene, acetone, chloroform,, diethyl ether and aromatic hydrocarbons
Formula Weight 78.13
Percent Purity 99.7%
Grade Pure
Chemical Name or Material Methyl sulfoxide, 99.7%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 200-664-3
RTECSNumber PV6210000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.