Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Octen-3-ol, 98%

Catalog No. AC129471000
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 3391-86-4 | C8H16O | 128.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3391-86-4
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00004589
InChI Key VSMOENVRRABVKN-UHFFFAOYSA-N
Synonym 1-octen-3-ol, vinyl amyl carbinol, 1-vinylhexanol, 3-hydroxy-1-octene, amyl vinyl carbinol, mushroom alcohol, matsutake alcohol, vinyl hexanol, pentyl vinyl carbinol, matsuica alcohol
PubChem CID 18827
ChEBI CHEBI:34118
IUPAC Name oct-1-en-3-ol
SMILES CCCCCC(C=C)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8370g/mL
Boiling Point 174.0°C
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4340 to 1.4420
Linear Formula CH3(CH2)4CH(OH)CH=CH2
Quantity 100 g
Specific Gravity 0.837
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in ethanol
Formula Weight 128.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Octen-3-ol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 222-226-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.