Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propargyl bromide, 80 wt.% solution in toluene, stabilized

Catalog No. AC131480010
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
AcroSeal™ Glass bottle
Glass bottle

CAS: 106-96-7 | C3H3Br | 118.961 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propargyl bromide
Color White
CAS 106-96-7, 108-88-3
Boiling Point 88.0°C to 90.0°C
Molecular Formula C3H3Br
Linear Formula HC≡CCH2Br
Fieser 06,493; 10,332; 06,495
Density 1.3800g/mL
Synonym propargyl bromide, 3-bromopropyne, 3-bromo-1-propyne, 1-propyne, 3-bromo, 2-propynyl bromide, propynyl bromide, 1-bromo-2-propyne, propyne, 3-bromo, gamma-bromoallylene, 1-brom-2-propin
Solubility Information Solubility in water: immiscible.
InChI Key YORCIIVHUBAYBQ-UHFFFAOYSA-N
SMILES C#CCBr
IUPAC Name 3-bromoprop-1-yne
Molecular Weight (g/mol) 118.961
PubChem CID 7842
Formula Weight 118.96
Percent Purity 73 to 87% (propargyl bromide) (GC) 13 to 27% (toluene) (GC)
Packaging AcroSeal™ Glass bottle
Refractive Index 1.4900 to 1.4960
Specific Gravity 1.38
Quantity 1 L
Flash Point 4°C
Physical Form Powder
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.