Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Rhodamine B, pure

Catalog No. AC132310025
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
Catalog No. Quantity Packaging
AC132310025 2.5 kg Plastic drum
AC132311000 100 g Glass bottle
AC132315000 500 g Glass bottle
3 options

Catalog No. AC132310025

Supplier: Thermo Scientific Chemicals 132310025

Only null left
Add to Cart

CAS: 81-88-9 | C28H31ClN2O3 | 479.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-88-9
Molecular Formula C28H31ClN2O3
Molecular Weight (g/mol) 479.02
MDL Number MFCD00011931
InChI Key PYWVYCXTNDRMGF-UHFFFAOYSA-N
Synonym Basic Violet 10, C.I. 45170
PubChem CID 6694
ChEBI CHEBI:52334
IUPAC Name [9-(2-carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium;chloride
SMILES [Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC

Specifications

Absorbance 1700 (at lambda min.)
Melting Point 210.0°C to 211.0°C
Absorption Ratio 1.1 to 1.6
Beilstein 19, V,8, 669
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in hcl and naoh, soluble in alcohol
Infrared Spectrum Authentic
Lambda Max 549 to 558nm (in 50:50 IMS:water)
Formula Weight 479.01
Loss on Drying 10% max. (105°C)
Grade Pure
Packaging Plastic drum
Color Green
Quantity 2.5 kg
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Rhodamine B
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Suspected of causing genetic defects.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.