Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nitric acid, extra pure, 60% solution in water, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless to Yellow |
| Melting Point | -42.0°C |
| Boiling Point | 122.0°C |
| CAS Min % | 59.0, 37.0 |
| CAS Max % | 63.0, 41.0 |
| Density | 1.3300g/mL |
| Packaging | Plastic bottle |
| Specific Gravity | 1.33 |
| Quantity | 1 L |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. May be corrosive to metals. Corrosive to the respiratory tract. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction