Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific Chemicals L(-)-Tryptophan, 99%
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
Packaging:
Glass Bottle
Plastic Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- L(-)-Tryptophan is a nonpolar aromatic compound and essential amino acid required for protein synthesis
- It is a biochemical precursor of niacin, indole alkaloids, and serotonin
Applications
- L-Tryptophan acts as a natural dietary supplement and has laboratory applications in the research of antidepressants, anxiolytics, and sleep aids
- It is a precursor of metabolites that are important in the laboratory study of mammalian physiology, including gastrointestinal functions, immunity, metabolism, and the nervous system
Specifications
Specifications
| Melting Point | 280.0°C to 285.0°C |
| Color | White to White-Yellow |
| Infrared Spectrum | Authentic |
| Loss on Drying | 0.5% max. (1 g, 105°C) |
| Assay Percent Range | 99% |
| Packaging | Plastic Bottle |
| Specific Rotation | − 31.10 |
| Beilstein | 22, 546 |
| Merck Index | 15, 9977 |
| Solubility Information | Solubility in water: 11.4g/L (25°C). Other solubilities: soluble in alkali hydroxides and hot alcohol, insoluble in chloroform |
| Show More |
Safety and Handling
| EINECSNumber | 200-795-6 |
| RTECSNumber | YN6130000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction