Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

delta-Valerolactam, 99%

Catalog No. AC140780250
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 675-20-7 | C5H9NO | 99.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 675-20-7
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00006037
InChI Key XUWHAWMETYGRKB-UHFFFAOYSA-N
Synonym 2-piperidone, 2-piperidinone, delta-valerolactam, valerolactim, 5-pentanolactam, piperidinone, piperidon, piperidone, alpha-piperidone, piperidone-2
PubChem CID 12665
ChEBI CHEBI:77761
IUPAC Name piperidin-2-one
SMILES O=C1CCCCN1

Specifications

Melting Point 35°C to 39°C
Color Brown to White
Density 1.0730g/mL
Boiling Point 256°C to 262°C
Flash Point 147°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Beilstein 21, 238
Specific Gravity 1.073
Solubility Information Solubility in water: 291g/L (25°C). Other solubilities: soluble in most common organic solvents,soluble in ethanol,ether
Viscosity 13.5 mPa.s (50°C)
Formula Weight 99.13
Percent Purity 99%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material δ-Valerolactam, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-622-9
RTECSNumber TO0110000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.