Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene glycol, 99+%, extra pure

Catalog No. AC146750250
Click to view available options
Quantity:
1 L
25 L
Packaging:
Glass Bottle
Plastic Drum

Catalog No. AC146750250

Supplier: Thermo Scientific Chemicals 146750250

Only null left
Add to Cart

CAS: 107-21-1 | C2H6O2 | 62.068 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
MDL Number MFCD00002885
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Melting Point 2°C to 3°C
Decomposition Information >500°C
Density 1.1130g/mL
Boiling Point 196.0°C to 198.0°C
Quantity 25 L
Flash Point 116°C (240.8°F)
Infrared Spectrum Authentic
Packaging Plastic Drum
Physical Form Viscous Liquid
Refractive Index 1.4300 to 1.4330 (20°C, 589nm)
Assay Percent Range 99+%
Linear Formula HOCH2CH2OH
Vapor Pressure Vapor Pressure: 0.12mmHg at 20°C
Beilstein 01, 465
Water 0.1% max. (Coulometric)
Fieser 01,375; 05,296; 09,217; 15,156
Merck Index 15, 3852
Formula Weight 62.06
Percent Purity ≥99.0%
Grade Extra Pure
Chemical Name or Material Ethylene glycol, 99+%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-473-3
RTECSNumber KW2975000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.