Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Methyl Orange, pure, indicator

Catalog No. AC151421000
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle

Catalog No. AC151421000

Supplier: Thermo Scientific Chemicals 151421000

Only null left
Add to Cart

CAS: 547-58-0 | C14H14N3NaO3S | 327.334 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 547-58-0
Molecular Formula C14H14N3NaO3S
Molecular Weight (g/mol) 327.334
MDL Number MFCD00007502
InChI Key STZCRXQWRGQSJD-UHFFFAOYSA-M
Synonym 4-[4-(Dimethylamino)phenylazo]benzenesulfonic acid, sodium salt, Acid Orange 52, C.I. 13025, MO, Helianthise
PubChem CID 23673835
IUPAC Name sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]

Specifications

Melting Point >300.0°C
Beilstein 16, 331
Merck Index 15, 6178
Solubility Information Solubility in water: soluble in hot water. Other solubilities: practically insoluble in alcohol
Infrared Spectrum Authentic
Lambda Max 461 to 469nm (in 0.01N NaOH)
Formula Weight 327.32
Grade Pure
Packaging Glass bottle
Visual Transition Interval from pH 3.2 (pink or red)to pH 4.4 (yellow-orange)
Color Orange
Quantity 100 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Methyl Orange
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-925-3
RTECSNumber DB6327000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.