Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chlorobutane, 99+%, pure

Catalog No. AC154630025
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 109-69-3 | C4H9Cl | 92.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-69-3
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.57
MDL Number MFCD00001009
InChI Key VFWCMGCRMGJXDK-UHFFFAOYSA-N
Synonym butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
PubChem CID 8005
IUPAC Name 1-chlorobutane
SMILES CCCCCl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -123.0°C
Color Colorless
Density 0.8800g/mL
Boiling Point 77.0°C to 78.0°C
Flash Point −12°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4014 to 1.4034
Linear Formula CH3(CH2)3Cl
Quantity 2.5 L
Beilstein 01, 118
Specific Gravity 0.88
Merck Index 15, 1561
Solubility Information Solubility in water: 0.5g/L (20°C). Other solubilities: miscible with alcohol and ether
Viscosity 0.45 mPa.s (20°C)
Formula Weight 92.57
Percent Purity 99+%
Grade Pure
Physical Form Liquid
Chemical Name or Material 1-Chlorobutane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o
EINECSNumber 203-696-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.