Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,10-Phenanthroline, 99+%

Catalog No. AC157531000
Click to view available options
Quantity:
5 g
25 g
100 g
500 g

CAS: 66-71-7 | C12H8N2 | 180.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-71-7
Molecular Formula C12H8N2
Molecular Weight (g/mol) 180.21
MDL Number MFCD00011678
InChI Key DGEZNRSVGBDHLK-UHFFFAOYSA-N
Synonym o-phenanthroline, 4,5-diazaphenanthrene, orthophenanthroline, phenanthroline, 1,10-o-phenanthroline, phen, 2-phenanthroline, beta-phenanthroline, anhydrous, .beta.-phenanthroline
PubChem CID 1318
ChEBI CHEBI:44975
IUPAC Name 1,10-phenanthroline
SMILES C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 117.0°C to 120.0°C
Color Pink or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 100 g
Beilstein 23,227
Merck Index 15,7328
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, acetone, ether and benzene
Formula Weight 180.21
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material 1, 10-Phenanthroline
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-629-2
RTECSNumber SF8300000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.