Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Pentanol, 99%, pure
Click to view available options
Quantity:
25 mL
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | -78.0°C |
Density | 0.8110g/mL |
Boiling Point | 137.0°C to 139.0°C |
Flash Point | 49°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Metal drum |
Refractive Index | 1.4083 to 1.4103 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Causes skin irritation. May cause respiratory irritation. Harmful if inhaled. Flammable liquid and vapor. Causes serious eye irritation. |
Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face,hands and any exposed skin thoroughly after handling. Wear protective glove |
EINECSNumber | 200-752-1 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction