Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Ouabain octahydrate, 96%

Catalog No. AC161730050
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Packaging:
Glass bottle

Ouabain octahydrate, CAS # 11018-89-6, (synonym: gamma-Strophanthin), is a plant-derived organic compound that inhibits the Na+/K+ ATPase pump present in all animal cell membranes. | CAS: 11018-89-6 | C29H44O12·8H2O | 728.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• Ouabain octahydrate is a water-soluble, light-sensitive powder that is stable at ambient temperatures
• It is a glycoside comprised of rhamnose and ouabagenin, and is derived from the ripe seeds of Strophanthus gratus and the bark of Acokanthera ouabaio

Applications

• Ouabain octahydrate works by binding the Na+/K+ ATPase pump, thus disrupting its function, and leading to an increase in intracellular Na+.
• The inhibitory action of ouabain octahydrate on the Na+/K+ pump in cardiac cells makes it a pharmaceutical cardioactive agent
• Ouabain octahydrate has also been found to induce cytotoxic effects through a steroidal signal transduction pathway, in laboratory preparations

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11018-89-6
Molecular Formula C29H44O12·8H2O
Molecular Weight (g/mol) 728.78
MDL Number MFCD00149240
InChI Key TYBARJRCFHUHSN-DMJRSANLSA-N
Synonym ouabain octahydrate, strophantine octahydrate, quabain octahydrate, ouabain usp, penta a phenanthren-17-yl-2h-furan-5-one;octahydrate, gamma-strophanthin, 3-1r,3s,5s,8r,9s,10r,11r,13r,14s,17r-1,5,11,14-tetrahydroxy-10-hydroxymethyl-13-methyl-3-2r,3r,4r,5r,6s-3,4,5-trihydroxy-6-methyloxan-2-yl oxy-2,3,4,6,7,8,9,11,12,15,16,17-dodecahydro-1h-cyclo, prestwick_370, acocantherine;g-strophanthin
PubChem CID 6364534
IUPAC Name 3-[(1R,3S,5S,8R,9S,10R,11R,13R,14S,17R)-1,5,11,14-tetrahydroxy-10-(hydroxymethyl)-13-methyl-3-[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy-2,3,4,6,7,8,9,11,12,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]-2H-furan-5-one;octahydrate
SMILES CC1C(C(C(C(O1)OC2CC(C3(C4C(CCC3(C2)O)C5(CCC(C5(CC4O)C)C6=CC(=O)OC6)O)CO)O)O)O)O.O.O.O.O.O.O.O.O

Specifications

Melting Point 260.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 15, V,5, 123
Merck Index 14, 6901
Specific Rotation Condition − 33.00 (20.00°C c=1,H2O,on dry subs.)
Specific Rotation − 33.00
Solubility Information (1% in water) Clear colorless
Formula Weight 728.78
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material Ouabain octahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Get medical attention/advice if you feel unwell.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.