Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Betaine hydrochloride, 99%, extra pure, specified according to the requirements of USP

Catalog No. AC164550010
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle

CAS: 590-46-5 | C5H12ClNO2 | 153.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-46-5
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00011903
InChI Key HOPSCVCBEOCPJZ-UHFFFAOYSA-N
Synonym betaine hydrochloride, betaine chloride, pluchine, acidogeno, acipepsol, achylin, acidin, acinorm, acidol, acidine
PubChem CID 11545
IUPAC Name carboxymethyl(trimethyl)azanium;chloride
SMILES [Cl-].C[N+](C)(C)CC(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 241.0°C
Color Colorless to White
pH 0.8 to 1.2 (1:4)
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula (CH3)3NCH2CO2H·Cl
Quantity 1 kg
Merck Index 15, 1181
Solubility Information Solubility in water: 647g/L (25°C). Other solubilities: soluble in ethanol,practically insoluble in chloroform and ether
Formula Weight 153.61
Percent Purity 99%
Grade Extra Pure
Physical Form Crystals or Powder
Chemical Name or Material βine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 209-683-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.