Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific Chemicals Acrylamide, 98.5%, extra pure
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Acrylamide is an organic amide compound mainly used to as a monomer in the synthesis of polyacrylamide
- Acrylamide is formed in foods containing carbohydrates in its composition, mainly when these foods are fried, grilled or baked
Applications
- In laboratory experiments acrylamide participates in chemical reactions as an intermediate in the preparation of N-methylol acrylamide and N-butoxyacrylamide compounds
- For molecular usage, this compound can be used as a carrier and is involved in the precipitation of small amounts of DNA
- In molecular experiments it is used in gel electrophoresis, including western blot, and SDS-PAGE, to separate proteins and macromolecules based on their size and charge

Specifications
Specifications
Physical Form | Crystals or Crystalline Powder |
Boiling Point | 125.0°C (25.0 mmHg) |
Melting Point | 82.0°C to 86.0°C |
Density | 1.1220g/mL |
Color | White |
Quantity | 2.5 kg |
Flash Point | 138°C |
Infrared Spectrum | Authentic |
Packaging | Plastic Bottle |
Assay Percent Range | 98.5% |
Show More |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS P Statement Causes skin irritation. Causes serious eye irritation. Toxic if swallowed. May cause an allergic skin reaction. Harmful if inhaled. May cause cancer. May cause genetic defects. Causes damage to organs through prolonged or repeated exposure. Suspected of damaging fertility. Harmful in contact with skin. |
Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Gently wash with plenty of soap and water. Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. Do not breathe dust/fume/gas/mist/vapors/spray. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction