Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Methyltetrahydrofuran, 99+%, pure, stabilized
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Metal Drum
Steel drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -136.0°C |
| Color | Colorless |
| Density | 0.8600g/mL |
| Boiling Point | 78.0°C to 80.0°C |
| Quantity | 2.5 kg |
| Infrared Spectrum | Authentic |
| Packaging | Steel drum |
| Physical Form | Liquid |
| Refractive Index | 1.4046 to 1.4066 |
| Assay Percent Range | 99% min. (GC) |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | GHS Signal Word: Danger |
| missing translation for 'healthHazard2' | GHS H Statement Highly flammable liquid and vapor. Harmful if swallowed. Causes skin irritation. Causes serious eye damage. May form explosive peroxides. |
| missing translation for 'healthHazard3' | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Wear protective g |
| EINECSNumber | 202-507-4 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction