Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iron(III) chloride, 98%, pure, anhydrous
Click to view available options
Quantity:
5 g
1 kg
2.5 kg
10 kg
25 kg
Packaging:
Fibre drum
Glass Bottle
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid

Specifications
Specifications
Color | Gray to Black |
Physical Form | Powder |
Quantity | 2.5 kg |
Assay Percent Range | 97.5 to 102.5% (Iodometry) |
Linear Formula | FeCl3 |
Fieser | 01,392; 02,199; 04,236; 05,307; 06,259; 07,153; 08,228; 09,222; 10,185; 11,237; 12,1; 13,133; 14,164; 16,167; 17,138 |
Merck Index | 15,4046 |
Solubility Information | Solubility in water: 480g/L (20°C). Other solubilities: readily soluble in alcohol, ether and acetone, slightly soluble in cs2, practically insoluble in ethyl acetate |
Formula Weight | 162.21 |
Percent Purity | 98% |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Harmful if swallowed. Causes serious eye damage. Causes skin irritation. May cause an allergic skin reaction. |
Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present |
EINECSNumber | 231-729-4 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction