Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-2-buten-1-ol, 99%

Catalog No. AC171921000
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 556-82-1 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-82-1
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00002916
InChI Key ASUAYTHWZCLXAN-UHFFFAOYSA-N
Synonym 3-methyl-2-buten-1-ol, prenol, 3,3-dimethylallyl alcohol, 2-buten-1-ol, 3-methyl, prenyl alcohol, 3-methyl-2-butenol, dimethylallyl alcohol, 3-methyl-2-butenyl alcohol, 3-methylcrotyl alcohol, 3-methyl-2-butene-1-ol
PubChem CID 11173
ChEBI CHEBI:16019
IUPAC Name 3-methylbut-2-en-1-ol
SMILES CC(C)=CCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8400g/mL
Boiling Point 140.0°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C=CHCH2OH
Refractive Index 1.4420 to 1.4440
Quantity 100 g
Beilstein 01, 444
Specific Gravity 0.84
Solubility Information Solubility in water: 170g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 86.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-buten-1-ol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
EINECSNumber 209-141-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.