Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals HEPES, 99%, for biochemistry

Catalog No. AC172570050
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
Synonym 2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid
PubChem CID 23831
ChEBI CHEBI:42334
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Absorbance 0.08 max. at 260nm, 0.05 max. at 280nm, 0.08 max. at 250nm, (5% in water) 1 cm cell, (5% in water) 1 cm cell at 0nm
Absorbance Conditions (5% aq. soln.) (1cm cell)
Melting Point 234.0°C to 238.0°C
Color (5% in water) clear, colorless
Decomposition Information 238°C
Physical Form Liquid or Solid
Quantity 5 kg
Assay Percent Range 99%
Infrared Spectrum Authentic
Loss on Drying 1 % max. (1 g, 105°C, 3 hrs)
Packaging Plastic drum
Beilstein 23, V,2, 379
Merck Index 15, 4689
Solubility Information Solubility in water: 704g/L (20°C).
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
Formula Weight 238.3
Percent Purity 99%
Grade Biochemical
Chemical Name or Material HEPES
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.