Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals PIPES, 98.5+%, for biochemistry

Catalog No. AC172615000
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 5625-37-6 | C8H18N2O6S2 | 302.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5625-37-6
Molecular Formula C8H18N2O6S2
Molecular Weight (g/mol) 302.36
InChI Key IHPYMWDTONKSCO-UHFFFAOYSA-N
Synonym 1, 4-Piperazinebis(ethanesulfonic acid)
PubChem CID 79723
ChEBI CHEBI:44933
IUPAC Name 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
SMILES C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O

Specifications

Absorbance (0.1M in 0.1M NaOH) (1 cm cell), (0.1M in 0.1M NaOH) (1 cm cell) at 0nm, 0.04 max. at 280nm, 0.05 max. at 260nm
Absorbance Conditions (0.1M in 0.1M NaOH) (1cm cell)
Color White
Physical Form Micro-crystalline Powder
Quantity 500 g
Assay Percent Range 98.5+%
Infrared Spectrum Authentic
Loss on Drying 0.5% max.
Packaging Plastic bottle
Formula Weight 302.35
Percent Purity ≥98.50%
Grade Biochemical
Chemical Name or Material PIPES
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.