Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
tert-Butyl hydroperoxide, 70% Solution in water
Click to view available options
Quantity:
5 g
250 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -2.8°C |
| Color | Colorless |
| Density | 0.9400g/mL |
| Boiling Point | 37.0°C (15.0 mmHg) |
| Flash Point | 43°C |
| Packaging | Plastic bottle |
| Refractive Index | 1.3860 to 1.3880 |
| Linear Formula | (CH3)3COOH |
| Quantity | 500 g |
| Beilstein | 01,IV,1616 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Toxic in contact with skin. Causes severe skin burns and eye damage. May cause an allergic skin reaction. Fatal if inhaled. Toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye p |
| EINECSNumber | 200-915-7 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction