Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
m-Xylene, 99+%, extra pure
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -48.0°C |
Color | Colorless |
CAS Min % | 99.0 |
CAS Max % | 100.0 |
Density | 0.8640g/mL |
Boiling Point | 139.0°C |
Quantity | 2.5 L |
Flash Point | 25°C |
Infrared Spectrum | Authentic |
Packaging | Glass Bottle |
Show More |
Safety and Handling
Hazard Category | Warning |
Hazard Statement | GHS H Statement: Causes skin irritation. Harmful in contact with skin. Harmful if inhaled. Flammable liquid and vapor. |
Precautionary Statement | GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction