Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2,6-Dichloro-3-nitropyridine, 90%, Tech.
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS Min % | 88.0 |
CAS Max % | 100.0 |
Melting Point | 58°C to 68°C |
Color | Colorless to Yellow |
Flash Point | >110°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 88% min. (GC) |
Packaging | Glass bottle |
Linear Formula | Cl2(C5H2N)NO2 |
Quantity | 50 g |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement May cause an allergic skin reaction. Causes serious eye irritation. Causes skin irritation. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position com |
EINECSNumber | 240-151-1 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction