Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cesium hydroxide monohydrate, 99.95%, (trace metal basis), for analysis
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 205.0°C to 208.0°C |
Color | Blue-Red to Green |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 99.95% |
Linear Formula | CsOH·H2O |
Merck Index | 10, 1970 |
Solubility Information | Solubility in water: soluble with exothermic reaction. Other solubilities: soluble in alcohol (exothermic) |
IUPAC Name | cesium;hydroxide;hydrate |
Formula Weight | 167.93 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Harmful if swallowed. Causes severe skin burns and eye damage. Suspected of damaging fertility or the unborn child. |
Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsi |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction