Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lutetium(III) oxide, 99.99%, (trace metal basis), -325 mesh

Catalog No. AC193400010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

Catalog No. AC193400010

Supplier: Thermo Scientific Chemicals 193400010

Only null left
Add to Cart

CAS: 12032-20-1 | Lu2O3 | 397.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12032-20-1
Molecular Formula Lu2O3
Molecular Weight (g/mol) 397.93
MDL Number MFCD00011100
InChI Key UGBIHFMRUDAMBY-UHFFFAOYSA-N
Synonym lutetium 3+ ; oxygen 2-, lutetium oxide, lutetium iii oxide trace rare earth metals basis 1g, cid159406, einecs 234-764-3, lu2o3, lutetium 3+
IUPAC Name dilutetium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Lu+3].[Lu+3]

Specifications

Color White
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.94% min
Linear Formula Lu2O3
Merck Index 14, 5615
Solubility Information Solubility in water: insoluble
Formula Weight 397.94
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 9.42
Density 9.4200g/mL
Chemical Name or Material Lutetium(III) oxide, 99.99%, -325 mesh
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 234-764-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.