Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Palladium(II) chloride, 59% Pd
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 678.0°C to 680.0°C |
Color | Red to Brown |
Physical Form | Powder |
Quantity | 25 g |
Linear Formula | PdCl2 |
Fieser | 01,782; 02,303; 04,369; 05,500; 06,447; 07,277; 08,384; 09,352; 10,300; 11,393; 12,371; 13,9; 15,248; 16,268 |
Merck Index | 15,7091 |
Solubility Information | Solubility in water: 31.99g/L. Other solubilities: soluble in hydrochloric acid,hydrobromic acid,soluble in alcohol and acetone |
IUPAC Name | dichloropalladium |
Formula Weight | 177.31 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement May be corrosive to metals. May cause an allergic skin reaction. Causes serious eye damage. Harmful if swallowed. Very toxic to aquatic life with long lasting effects. |
Precautionary Statement | GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective |
EINECSNumber | 231-596-2 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction