Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium sulfate, 99%, extra pure, anhydrous

Catalog No. AC196640251
Click to view available options
Quantity:
25 g
1 kg
2.5 kg
5 kg
25 kg
Packaging:
Fibre drum
Glass Bottle
Plastic bottle

CAS: 7757-82-6 | Na2O4S | 142.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Boiling Point >1700.0°C
Melting Point 884.0°C
Color White
Physical Form Crystalline Powder
Quantity 25 kg
Assay Percent Range 98.5% min
Linear Formula Na2SO4
Fieser 01,1103
Merck Index 15, 8809
Solubility Information Solubility in water: 185g/L. Other solubilities: soluble in glycerol and hydrogen iodide, insoluble in alcohol
IUPAC Name disodium sulfate
Formula Weight 142.04
Percent Purity 99%
Grade Extra Pure
Packaging Fibre drum
Density 2.68
Chemical Name or Material Sodium sulfate
Show More Show Less
EINECSNumber 231-820-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.