Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ruthenium(III) chloride, 99.9%, (trace metal basis), anhydrous
Click to view available options
Quantity:
2 g
10 g
50 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
Specifications
Specifications
| Melting Point | >500.0°C |
| Color | Black |
| Physical Form | Crystals or Powder |
| Quantity | 10 g |
| Assay Percent Range | 48 to 50% (Ru) |
| Linear Formula | RuCl3 |
| Fieser | 04,421; 08,437; 10,343; 13,268; 14,271; 15,280; 16,292 |
| Merck Index | 15,8436 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: decomposes in hot water, slightly soluble in alcohol |
| Formula Weight | 207.43 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful to aquatic life with long lasting effects. Causes severe skin burns and eye damage. May be corrosive to metals. Harmful if swallowed. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with wat |
| EINECSNumber | 233-167-5 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction