Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) nitrate nonahydrate, 99+%, for analysis

Catalog No. AC202492500
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog No. Quantity Packaging
AC202492500 250 g Plastic bottle
AC202490010 1 kg Plastic bottle
AC202490050 5 kg Plastic bottle
3 options

Catalog No. AC202492500

Supplier: Thermo Scientific Chemicals 202492500

Only null left
Add to Cart

CAS: 7782-61-8 | FeH18N3O18 | 403.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-61-8
Molecular Formula FeH18N3O18
Molecular Weight (g/mol) 403.99
MDL Number MFCD00149708
InChI Key SZQUEWJRBJDHSM-UHFFFAOYSA-N
Synonym ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
PubChem CID 16211566
SMILES O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 47.0°C
Color Purple
Physical Form Slightly Adhering Crystalline Powder
Quantity 250 g
Assay Percent Range 99% min. (Iodometry)
Linear Formula Fe(NO3)3·9H2O
Merck Index 15,4054
Solubility Information Solubility in water: soluble. Other solubilities: very soluble in ethanol, acetone
IUPAC Name iron(3+) nonahydrate trinitrate
Formula Weight 404
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 1.68
Density 1.6800g/mL
Chemical Name or Material Iron(III) nitrate nonahydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cauti

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.