Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lithium iodide, 99%, pure, anhydrous
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1171.0°C |
| Melting Point | 446.0°C |
| Physical Form | Powder, Crystals and/or Granules |
| Quantity | 250 g |
| Assay Percent Range | 98.5% min. (on anhydrous substance) (Argentometry) |
| Linear Formula | LiI |
| Fieser | 01,615; 04,304; 05,410; 07,208; 09,283; 10,245; 11,300; 12,282; 15,185; 16,197; 17,85 |
| Merck Index | 15, 5591 |
| Solubility Information | Solubility in water: 165 g/100 mL (20°C). Other solubilities: soluble in alcohol, freely soluble in acetone |
| Formula Weight | 133.85 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement May cause respiratory irritation. Causes serious eye irritation. Causes skin irritation. |
| Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at |
| EINECSNumber | 233-822-5 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction