Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Molybdenum(VI) oxide, 99+%
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1155.0°C |
| Melting Point | 795.0°C |
| Color | Green-Yellow |
| CAS Min % | 99.0 |
| CAS Max % | 100.0 |
| Physical Form | Powder |
| Quantity | 100 g |
| Assay Percent Range | 99% min |
| Linear Formula | MoO3 |
| Fieser | 04,203; 13,279; 16,227 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Suspected of causing cancer. May cause respiratory irritation. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Wear protective gloves/protective clot |
| EINECSNumber | 215-204-7 |
| RTECSNumber | QA4725000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction