Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium chlorate, 99%, extra pure, Thermo Scientific Chemicals

Catalog No. AC208860010
Click to view available options
Quantity:
1 kg
Packaging:
Plastic bottle

Catalog No. AC208860010

Supplier: Thermo Scientific Chemicals 208860010

Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3811-04-9
Molecular Formula ClKO3
Molecular Weight (g/mol) 122.55
MDL Number MFCD00011361
InChI Key VKJKEPKFPUWCAS-UHFFFAOYSA-M
Synonym potassium chlorate, fekabit, potcrate, kaliumchlorat, anforstan, berthollet salt, salt of tarter, berthollet's salt, chlorate of potash, chloric acid, potassium salt
PubChem CID 6426889
IUPAC Name potassium;chlorate
SMILES [O-]Cl(=O)=O.[K+]

Specifications

Melting Point 356.0°C to 368.0°C
Color White
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 98.5% min. (KClO3)
Linear Formula KClO3
Fieser 01,927
Merck Index 15, 7741
Solubility Information Solubility in water: 73g/L (20°C). Other solubilities: 555g/L hot water (100°C), soluble in alkalies, 1g/50mL glycerol, practically insoluble in alcohol, insoluble in ether
Formula Weight 122.55
Percent Purity 99%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Potassium chlorate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Harmful if inhaled.
May cause fire or explosion; strong oxidizer.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major fire and large quantities: Evacuate area.
Fight fire remotely due to the ris
EINECSNumber 223-289-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.