Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium nitrate, 99+%, extra pure

Catalog No. AC211320010
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle

CAS: 7790-69-4 | LiNO3 | 68.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-69-4
Molecular Formula LiNO3
Molecular Weight (g/mol) 68.94
MDL Number MFCD00011094
InChI Key IIPYXGDZVMZOAP-UHFFFAOYSA-N
Synonym lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure
PubChem CID 10129889
ChEBI CHEBI:63315
SMILES [Li+].[O-][N+]([O-])=O

Specifications

Boiling Point 600.0°C
Melting Point 251.0°C
Color White
Physical Form Adhering Crystalline Powder
Quantity 1 kg
Assay Percent Range 99+%
Linear Formula LiNO3
Merck Index 15, 5592
Solubility Information Solubility in water: 90g/L (28°C). Other solubilities: soluble in alcohol
IUPAC Name lithium;nitrate
Formula Weight 68.95
Percent Purity ≥99%
Grade Extra Pure
Packaging Plastic bottle
Specific Gravity 2.38
Density 2.3800g/mL
Chemical Name or Material Lithium nitrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
May intensify fire; oxidizer.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
EINECSNumber 232-218-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.