Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lithium nitrate, 99+%, extra pure
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 600.0°C |
Melting Point | 251.0°C |
Color | White |
Physical Form | Adhering Crystalline Powder |
Quantity | 5 kg |
Assay Percent Range | 99+% |
Linear Formula | LiNO3 |
Merck Index | 15, 5592 |
Solubility Information | Solubility in water: 90g/L (28°C). Other solubilities: soluble in alcohol |
IUPAC Name | lithium(1+) nitrate |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Harmful if swallowed. May intensify fire; oxidizer. Causes serious eye irritation. |
Precautionary Statement | GHS P Statement IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF IN EYES: Rinse cautiously with water for several minutes. Remove conta |
EINECSNumber | 232-218-9 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction