Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium peroxomonosulfate, extra pure, min. 4.5% active oxygen
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Specifications
| Color | White |
| Physical Form | Granular Crystalline Powder |
| Quantity | 1 kg |
| Linear Formula | KHSO4·K2SO4 |
| Fieser | 01,952; 12,413; 16,285 |
| Solubility Information | Solubility in water: 298g/L (20°C). |
| Formula Weight | 614.78 |
| Percent Purity | ≥4.5% (active oxygen) |
| Grade | Extra Pure |
| Packaging | Glass bottle |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Causes severe skin burns and eye damage. Harmful to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with wat |
| EINECSNumber | 274-778-7 |
| TSCA | Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction