Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cobalt(II) chloride, 97%, anhydrous
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
| Boiling Point | 1049.0°C |
| Melting Point | 735.0°C |
| Color | Blue |
| Physical Form | Powder |
| Quantity | 100 g |
| Assay Percent Range | 97% |
| Linear Formula | CoCl2 |
| Fieser | 01,155; 06,127; 08,291; 10,101; 13,249; 14,99; 15,97 |
| Merck Index | 15, 2421 |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol and acetone |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause cancer by inhalation. Suspected of causing genetic defects. May damage fertility. Causes serious eye damage. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cau |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water. IF |
| EINECSNumber | 231-589-4 |
| RTECSNumber | GF9800000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction