Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triton™ X-100, Thermo Scientific™

Catalog No. AC215680025
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
Synonym Polyoxyethylene(10) octylphenyl ether
PubChem CID 5590
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Specifications

Boiling Point 270.0°C
Melting Point 6.0°C
Density 1.0670g/mL
Quantity 2.5 L
Flash Point 274°C
Packaging Glass Bottle
Refractive Index 1.4900 to 1.4920
Specific Gravity 1.067
Viscosity 240 mPa.s (25°C)
Merck Index 15, 6850
Solubility Information Solubility in water: soluble. Other solubilities: soluble in toluene, xylene, trichloroethylene,, ethylene glycol, ethyl ether, ethyl alcohol,, isopropyl alcohol, ethylene dichloride and most, other solvnets., insoluble in kerosene, miniral spirits.
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
Formula Weight 646.85
Chemical Name or Material Triton™ X-100
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Harmful to aquatic life with long lasting effects.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.