Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) chloride hexahydrate, 99+%, for analysis

Catalog No. AC217090025
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 10025-77-1 | Cl3FeH12O6 | 270.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid for analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-77-1
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
Synonym Ferric chloride hexahydrate
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Boiling Point 280.0°C to 285.0°C
Melting Point 37.0°C
Color Orange
Physical Form Adhering Crystalline Powder and Lumps
Quantity 2.5 kg
Assay Percent Range 99.0 to 102% (Iodometry)
Linear Formula FeCl3·6H2O
Fieser 01,390; 02,199; 03,145; 04,236; 05,307; 06,259; 07,153; 08,228; 09,222; 15,158
Merck Index 15, 4046
Solubility Information Solubility in water: 920g/L (20°C). Other solubilities: readily soluble in alcohol,acetone and ether
Formula Weight 270.29
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Iron(III) chloride hexahydrate, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with wa
RTECSNumber NO5425000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.