Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chromium(III) nitrate nonahydrate, 99%
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 60.0°C |
Color | Black to Purple |
pH | 2 to 3 (5% aq. soln.) |
Physical Form | Crystalline Chunks |
Quantity | 500 g |
Assay Percent Range | 98.5% min. (Iodometry) |
Molecular Formula | CrN3O9·9H2O |
Linear Formula | Cr(NO3)3·9H2O |
Merck Index | 14,2226 |
Solubility Information | Solubility in water: 1g/L (20°C). Other solubilities: soluble in alcohol |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Harmful if inhaled. Causes serious eye irritation. May intensify fire; oxidizer. May cause an allergic skin reaction. Toxic to aquatic life with long lasting effects. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Take any precaution to avoid mixing with combustibles. Call a POISON CE |
RTECSNumber | GB6300000 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction